Assay Details
| Gene Symbol: EDA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115900.4 | NP_191595.1 | 8 - 9 | 811 | 86 | |
| GenBank mRNA | AY039876.1 | - | 8 - 9 | 783 | 86 | |
| AY085156.1 | - | 8 - 9 | 766 | 86 | ||
| AY101514.1 | - | 7 - 8 | 592 | 86 | ||
More Information
Set Membership: |
|

EDA14
Genomic Map