Assay Details
| Gene Symbol: TTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340058.1 | NP_001327227.1 | 15 - 16 | 3221 | 76 | |
| NM_115938.6 | NP_191633.3 | 15 - 16 | 3481 | 76 | ||
| GenBank mRNA | AF486851.1 | - | 15 - 16 | 3285 | 76 | |
More Information
Set Membership: |
|

TTN1
Genomic Map