Assay Details
| Gene Symbol: TTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340058.1 | NP_001327227.1 | 4 - 5 | 987 | 94 | |
| NM_001340059.1 | NP_001327226.1 | 4 - 5 | 1090 | 94 | ||
| NM_115938.6 | NP_191633.3 | 4 - 5 | 1247 | 94 | ||
| GenBank mRNA | AF486851.1 | - | 4 - 5 | 1051 | 94 | |
More Information
Set Membership: |
|

TTN1
Genomic Map