Assay Details
| Gene Symbol: AT3G60980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115962.4 | NP_191657.1 | 1 | 1189 | 106 | |
| GenBank mRNA | AY081333.1 | - | 1 | 1161 | 106 | |
| BT002139.1 | - | 1 | 1094 | 106 | ||
More Information
Set Membership: |
|

AT3G60980
Genomic Map