Assay Details
| Gene Symbol: FUS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115978.4 | NP_567109.1 | 6 - 7 | 1520 | 134 | |
| GenBank mRNA | AF360295.1 | - | 6 - 7 | 1406 | 134 | |
More Information
Set Membership: |
|

FUS6
Genomic Map