Assay Details
| Gene Symbol: AT3G61290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115993.2 | NP_191688.1 | 3 - 4 | 778 | 91 | |
| GenBank mRNA | KJ138779.1 | - | 3 - 4 | 778 | 91 | |
More Information
Set Membership: |
|

AT3G61290
Genomic Map