Assay Details
| Gene Symbol: SKIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340087.1 | NP_001327045.1 | 1 - 2 | 931 | 114 | |
| NM_115999.5 | NP_567112.1 | 2 - 3 | 635 | 114 | ||
| GenBank mRNA | BT025245.1 | - | 1 - 2 | 487 | 114 | |
More Information
Set Membership: |
|

SKIP4
Genomic Map