Assay Details
| Gene Symbol: CYP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084509.1 | NP_001077978.1 | 1 - 2 | 116 | 64 | |
| NM_001336236.1 | NP_001318316.1 | 1 - 2 | 146 | 64 | ||
| NM_128550.4 | NP_180557.1 | 1 - 2 | 205 | 64 | ||
| GenBank mRNA | AY054593.1 | - | 1 - 2 | 108 | 64 | |
| BT000068.1 | - | 1 - 2 | 65 | 64 | ||
More Information
Set Membership: |
|

CYP5
Genomic Map