Assay Details
| Gene Symbol: CYP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084509.1 | NP_001077978.1 | 2 - 3 | 187 | 94 | |
| NM_001336236.1 | NP_001318316.1 | 2 - 3 | 217 | 94 | ||
| NM_128550.4 | NP_180557.1 | 2 - 3 | 276 | 94 | ||
| GenBank mRNA | AY054593.1 | - | 2 - 3 | 179 | 94 | |
| BT000068.1 | - | 2 - 3 | 136 | 94 | ||
More Information
Set Membership: |
|

CYP5
Genomic Map