Assay Details
| Gene Symbol: CYP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336236.1 | NP_001318316.1 | 5 - 6 | 471 | 103 | |
| NM_128550.4 | NP_180557.1 | 5 - 6 | 530 | 103 | ||
| GenBank mRNA | AY054593.1 | - | 5 - 6 | 433 | 103 | |
| BT000068.1 | - | 5 - 6 | 390 | 103 | ||
More Information
Set Membership: |
|

CYP5
Genomic Map