Assay Details
| Gene Symbol: AT2G29970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128551.4 | NP_565689.1 | 1 - 2 | 1550 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G29970| Gene Symbol: AT2G29970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128551.4 | NP_565689.1 | 1 - 2 | 1550 | 132 | |
Set Membership: |
|