Assay Details
| Gene Symbol: ASL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128568.2 | NP_565695.1 | 1 - 2 | 439 | 63 | |
| GenBank mRNA | AB164306.1 | - | 1 - 2 | 160 | 63 | |
| AB473838.1 | - | 1 - 2 | 221 | 63 | ||
More Information
Set Membership: |
|

ASL5
Genomic Map