Assay Details
| Gene Symbol: UGT87A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128569.4 | NP_180575.1 | 1 - 2 | 707 | 86 | |
| GenBank mRNA | AK226350.1 | - | 1 - 2 | 543 | 86 | |
| AY093176.1 | - | 1 - 2 | 557 | 86 | ||
| BT006597.1 | - | 1 - 2 | 491 | 86 | ||
| KJ138888.1 | - | 1 - 2 | 491 | 86 | ||
More Information
Set Membership: |
|

UGT87A2
Genomic Map