Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202710.1 | NP_001189639.1 | 8 - 9 | 1280 | 108 | |
| NM_128592.3 | NP_180598.1 | 8 - 9 | 1284 | 108 | ||
| GenBank mRNA | BT000465.1 | - | 8 - 9 | 1227 | 108 | |
| BT008877.1 | - | 8 - 9 | 1121 | 108 | ||
More Information
Set Membership: |
|

FC2
Genomic Map