Assay Details
| Gene Symbol: GILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343258.1 | NP_001330776.1 | 1 - 2 | 451 | 61 | |
| NM_121322.2 | NP_568286.1 | 1 - 2 | 468 | 61 | ||
| GenBank mRNA | AY034946.1 | - | 1 - 2 | 263 | 61 | |
| AY062988.1 | - | 1 - 2 | 198 | 61 | ||
| AY088302.1 | - | 1 - 2 | 398 | 61 | ||
More Information
Set Membership: |
|

GILP
Genomic Map