Assay Details
| Gene Symbol: GILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121322.2 | NP_568286.1 | 2 - 3 | 591 | 59 | |
| GenBank mRNA | AY034946.1 | - | 2 - 3 | 386 | 59 | |
| AY062988.1 | - | 2 - 3 | 321 | 59 | ||
| AY088302.1 | - | 2 - 3 | 521 | 59 | ||
More Information
Set Membership: |
|

GILP
Genomic Map