Assay Details
| Gene Symbol: AK-LYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121331.3 | NP_196832.1 | 1 - 2 | 605 | 109 | |
| GenBank mRNA | AY057674.1 | - | 1 - 2 | 479 | 109 | |
| BT000493.1 | - | 1 - 2 | 401 | 109 | ||
More Information
Set Membership: |
|

AK-LYS1
Genomic Map