Assay Details
| Gene Symbol: SFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121333.4 | NP_196834.3 | 7 - 8 | 1021 | 66 | |
| GenBank mRNA | AB194395.1 | - | 7 - 8 | 471 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFC| Gene Symbol: SFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121333.4 | NP_196834.3 | 7 - 8 | 1021 | 66 | |
| GenBank mRNA | AB194395.1 | - | 7 - 8 | 471 | 66 | |
Set Membership: |
|