Assay Details
| Gene Symbol: AT5G13560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121359.5 | NP_196860.2 | 13 - 14 | 1540 | 78 | |
| GenBank mRNA | AK117870.1 | - | 13 - 14 | 1449 | 78 | |
| BT005947.1 | - | 13 - 14 | 1357 | 78 | ||
More Information
Set Membership: |
|

AT5G13560
Genomic Map