Assay Details
| Gene Symbol: AGL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343293.1 | NP_001330207.1 | 1 - 2 | 339 | 71 | |
| NM_121382.3 | NP_196883.1 | 1 - 2 | 245 | 71 | ||
More Information
Set Membership: |
|

AGL15
Genomic Map