Assay Details
| Gene Symbol: TBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343296.1 | NP_001330208.1 | 1 - 2 | 218 | 130 | |
| NM_121385.5 | NP_196886.1 | 2 - 3 | 446 | 130 | ||
| GenBank mRNA | AY029195.1 | - | 2 - 3 | 371 | 130 | |
| BT046176.1 | - | 1 - 2 | 78 | 130 | ||
More Information
Set Membership: |
|

TBP1
Genomic Map