Assay Details
| Gene Symbol: TBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343296.1 | NP_001330208.1 | 3 - 4 | 1114 | 111 | |
| NM_121385.5 | NP_196886.1 | 4 - 5 | 1342 | 111 | ||
| GenBank mRNA | AY029195.1 | - | 4 - 5 | 1267 | 111 | |
| BT046176.1 | - | 3 - 4 | 974 | 111 | ||
More Information
Set Membership: |
|

TBP1
Genomic Map