Assay Details
| Gene Symbol: XTH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343297.1 | NP_001331376.1 | 1 - 2 | 1065 | 79 | |
| NM_001343298.1 | NP_001331375.1 | 2 - 3 | 788 | 79 | ||
| NM_121390.5 | NP_196891.1 | 2 - 3 | 634 | 79 | ||
| GenBank mRNA | AB026486.1 | - | 1 - 2 | 232 | 79 | |
| AK176228.1 | - | 1 - 2 | 212 | 79 | ||
More Information
Set Membership: |
|

XTH5
Genomic Map