Assay Details
| Gene Symbol: AT5G13900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121393.4 | NP_196894.1 | 1 - 2 | 396 | 81 | |
| GenBank mRNA | AK175242.1 | - | 1 - 2 | 335 | 81 | |
| BT015106.1 | - | 1 - 2 | 322 | 81 | ||
More Information
Set Membership: |
|

AT5G13900
Genomic Map