Assay Details
| Gene Symbol: SUVH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343303.1 | NP_001332445.1 | 9 - 10 | 1489 | 113 | |
| NM_121399.3 | NP_196900.1 | 10 - 11 | 1253 | 113 | ||
| GenBank mRNA | BT002313.1 | - | 9 - 10 | 1081 | 113 | |
More Information
Set Membership: |
|

SUVH4
Genomic Map