Assay Details
| Gene Symbol: SUVH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121399.3 | NP_196900.1 | 3 - 4 | 571 | 69 | |
| GenBank mRNA | BT002313.1 | - | 2 - 3 | 399 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUVH4| Gene Symbol: SUVH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121399.3 | NP_196900.1 | 3 - 4 | 571 | 69 | |
| GenBank mRNA | BT002313.1 | - | 2 - 3 | 399 | 69 | |
Set Membership: |
|