Assay Details
| Gene Symbol: MPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343319.1 | NP_001330562.1 | 1 - 2 | 324 | 96 | |
| NM_001343321.1 | NP_001330560.1 | 1 - 2 | 324 | 96 | ||
| NM_121422.4 | NP_568295.2 | 1 - 2 | 462 | 96 | ||
| GenBank mRNA | AK176026.1 | - | 1 - 2 | 222 | 96 | |
More Information
Set Membership: |
|

MPL1
Genomic Map