Assay Details
| Gene Symbol: MPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343318.1 | NP_001330561.1 | 5 - 6 | 675 | 78 | |
| NM_001343319.1 | NP_001330562.1 | 6 - 7 | 887 | 78 | ||
| NM_001343320.1 | NP_001330559.1 | 5 - 6 | 553 | 78 | ||
| NM_001343321.1 | NP_001330560.1 | 6 - 7 | 887 | 78 | ||
| NM_121422.4 | NP_568295.2 | 6 - 7 | 1025 | 78 | ||
| GenBank mRNA | AK176026.1 | - | 6 - 7 | 785 | 78 | |
More Information
Set Membership: |
|

MPL1
Genomic Map