Assay Details
| Gene Symbol: HEMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203378.1 | NP_001190307.1 | 10 - 11 | 729 | 140 | |
| NM_001343324.1 | NP_001331496.1 | 10 - 11 | 1133 | 140 | ||
| NM_001343325.1 | NP_001331495.1 | 11 - 12 | 1024 | 140 | ||
| NM_001343326.1 | NP_001331494.1 | 11 - 12 | 835 | 140 | ||
| NM_121426.5 | NP_196926.2 | 11 - 12 | 798 | 140 | ||
| GenBank mRNA | AY075665.1 | - | 11 - 12 | 773 | 140 | |
| AY101523.1 | - | 11 - 12 | 740 | 140 | ||
More Information
Set Membership: |
|

HEMG2
Genomic Map