Assay Details
| Gene Symbol: HEMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203378.1 | NP_001190307.1 | 12 - 13 | 979 | 111 | |
| NM_001343324.1 | NP_001331496.1 | 12 - 13 | 1383 | 111 | ||
| NM_001343325.1 | NP_001331495.1 | 13 - 14 | 1274 | 111 | ||
| NM_001343326.1 | NP_001331494.1 | 13 - 14 | 1085 | 111 | ||
| NM_121426.5 | NP_196926.2 | 13 - 14 | 1048 | 111 | ||
| GenBank mRNA | AY075665.1 | - | 13 - 14 | 1023 | 111 | |
| AY101523.1 | - | 13 - 14 | 990 | 111 | ||
More Information
Set Membership: |
|

HEMG2
Genomic Map