Assay Details
| Gene Symbol: HEMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343324.1 | NP_001331496.1 | 4 - 5 | 724 | 70 | |
| NM_001343325.1 | NP_001331495.1 | 5 - 6 | 615 | 70 | ||
| NM_001343326.1 | NP_001331494.1 | 5 - 6 | 426 | 70 | ||
| NM_121426.5 | NP_196926.2 | 5 - 6 | 389 | 70 | ||
| GenBank mRNA | AY075665.1 | - | 5 - 6 | 364 | 70 | |
| AY101523.1 | - | 5 - 6 | 331 | 70 | ||
More Information
Set Membership: |
|

HEMG2
Genomic Map