Assay Details
| Gene Symbol: HEMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203378.1 | NP_001190307.1 | 7 - 8 | 492 | 67 | |
| NM_001343324.1 | NP_001331496.1 | 7 - 8 | 896 | 67 | ||
| NM_001343325.1 | NP_001331495.1 | 8 - 9 | 787 | 67 | ||
| NM_001343326.1 | NP_001331494.1 | 8 - 9 | 598 | 67 | ||
| NM_121426.5 | NP_196926.2 | 8 - 9 | 561 | 67 | ||
| GenBank mRNA | AY075665.1 | - | 8 - 9 | 536 | 67 | |
| AY101523.1 | - | 8 - 9 | 503 | 67 | ||
More Information
Set Membership: |
|

HEMG2
Genomic Map