Assay Details
| Gene Symbol: AT5G14470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121451.5 | NP_196951.3 | 4 - 5 | 716 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G14470| Gene Symbol: AT5G14470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121451.5 | NP_196951.3 | 4 - 5 | 716 | 89 | |
Set Membership: |
|