Assay Details
| Gene Symbol: AT5G14580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121462.4 | NP_196962.1 | 14 - 15 | 2215 | 111 | |
| GenBank mRNA | Y14685.1 | - | 2137 | 111 | ||
More Information
Set Membership: |
|

AT5G14580
Genomic Map