Assay Details
| Gene Symbol: AT5G14580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121462.4 | NP_196962.1 | 8 - 9 | 1434 | 78 | |
| GenBank mRNA | Y14685.1 | - | 1356 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G14580| Gene Symbol: AT5G14580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121462.4 | NP_196962.1 | 8 - 9 | 1434 | 78 | |
| GenBank mRNA | Y14685.1 | - | 1356 | 78 | ||
Set Membership: |
|