Assay Details
| Gene Symbol: SK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343350.1 | NP_001329975.1 | 12 - 13 | 1255 | 80 | |
| NM_121468.4 | NP_196968.2 | 12 - 13 | 1459 | 80 | ||
| GenBank mRNA | AY065043.1 | - | 12 - 13 | 1367 | 80 | |
| BT010466.1 | - | 11 - 12 | 1154 | 80 | ||
More Information
Set Membership: |
|

SK13
Genomic Map