Assay Details
| Gene Symbol: SK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343350.1 | NP_001329975.1 | 4 - 5 | 371 | 110 | |
| NM_121468.4 | NP_196968.2 | 4 - 5 | 575 | 110 | ||
| GenBank mRNA | AY065043.1 | - | 4 - 5 | 483 | 110 | |
| BT010466.1 | - | 3 - 4 | 270 | 110 | ||
More Information
Set Membership: |
|

SK13
Genomic Map