Assay Details
| Gene Symbol: AT5G14790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121483.3 | NP_196983.1 | 2 - 3 | 532 | 148 | |
| GenBank mRNA | AK221795.1 | - | 2 - 3 | 468 | 148 | |
| AY088875.1 | - | 2 - 3 | 493 | 148 | ||
| AY093238.1 | - | 2 - 3 | 495 | 148 | ||
| BT001241.1 | - | 1 - 2 | 266 | 148 | ||
More Information
Set Membership: |
|

AT5G14790
Genomic Map