Assay Details
| Gene Symbol: AT5G14790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121483.3 | NP_196983.1 | 4 - 5 | 770 | 131 | |
| GenBank mRNA | AK221795.1 | - | 4 - 5 | 706 | 131 | |
| AY088875.1 | - | 4 - 5 | 731 | 131 | ||
| AY093238.1 | - | 4 - 5 | 733 | 131 | ||
| BT001241.1 | - | 3 - 4 | 504 | 131 | ||
More Information
Set Membership: |
|

AT5G14790
Genomic Map