Assay Details
| Gene Symbol: AT5G14790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121483.3 | NP_196983.1 | 5 - 6 | 969 | 100 | |
| GenBank mRNA | AK221795.1 | - | 5 - 6 | 905 | 100 | |
| AY088875.1 | - | 5 - 6 | 930 | 100 | ||
| AY093238.1 | - | 5 - 6 | 932 | 100 | ||
| BT001241.1 | - | 4 - 5 | 703 | 100 | ||
More Information
Set Membership: |
|

AT5G14790
Genomic Map