Assay Details
| Gene Symbol: DEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343379.1 | NP_001330250.1 | 1 - 2 | 82 | 108 | |
| NM_121500.4 | NP_197000.1 | 5 - 6 | 441 | 108 | ||
| GenBank mRNA | AB074531.1 | - | 5 - 6 | 330 | 108 | |
| AB493750.1 | - | 5 - 6 | 330 | 108 | ||
| AJ417835.1 | - | 5 - 6 | 348 | 108 | ||
More Information
Set Membership: |
|

DEL2
Genomic Map