Assay Details
| Gene Symbol: DEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343379.1 | NP_001330250.1 | 3 - 4 | 222 | 85 | |
| NM_121500.4 | NP_197000.1 | 7 - 8 | 581 | 85 | ||
| GenBank mRNA | AB074531.1 | - | 7 - 8 | 470 | 85 | |
| AB493750.1 | - | 7 - 8 | 470 | 85 | ||
| AJ417835.1 | - | 7 - 8 | 488 | 85 | ||
More Information
Set Membership: |
|

DEL2
Genomic Map