Assay Details
| Gene Symbol: DEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343379.1 | NP_001330250.1 | 5 - 6 | 361 | 90 | |
| NM_121500.4 | NP_197000.1 | 9 - 10 | 720 | 90 | ||
| GenBank mRNA | AB074531.1 | - | 9 - 10 | 609 | 90 | |
| AB493750.1 | - | 9 - 10 | 609 | 90 | ||
| AJ417835.1 | - | 9 - 10 | 627 | 90 | ||
More Information
Set Membership: |
|

DEL2
Genomic Map