Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203385.1 | NP_001190314.1 | 3 - 4 | 434 | 75 | |
| NM_121513.5 | NP_197013.1 | 3 - 4 | 404 | 75 | ||
| GenBank mRNA | AJ131391.1 | - | 3 - 4 | 274 | 75 | |
| AY063891.1 | - | 3 - 4 | 291 | 75 | ||
| AY122918.1 | - | 3 - 4 | 220 | 75 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map