Assay Details
| Gene Symbol: HB30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121525.4 | NP_197025.1 | 1 | 1242 | 75 | |
| GenBank mRNA | BT012195.1 | - | 1 | 724 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HB30| Gene Symbol: HB30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121525.4 | NP_197025.1 | 1 | 1242 | 75 | |
| GenBank mRNA | BT012195.1 | - | 1 | 724 | 75 | |
Set Membership: |
|