Assay Details
| Gene Symbol: CASP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121533.4 | NP_197033.1 | 1 - 2 | 305 | 65 | |
| GenBank mRNA | AK117342.1 | - | 1 - 2 | 305 | 65 | |
| BT003725.1 | - | 1 - 2 | 222 | 65 | ||
More Information
Set Membership: |
|

CASP5
Genomic Map