Assay Details
| Gene Symbol: CASP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121533.4 | NP_197033.1 | 2 - 3 | 428 | 87 | |
| GenBank mRNA | AK117342.1 | - | 2 - 3 | 428 | 87 | |
| BT003725.1 | - | 2 - 3 | 345 | 87 | ||
More Information
Set Membership: |
|

CASP5
Genomic Map