Assay Details
| Gene Symbol: SPX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343398.1 | NP_001330175.1 | 1 - 2 | 556 | 154 | |
| NM_121537.3 | NP_568312.1 | 1 - 2 | 508 | 154 | ||
| GenBank mRNA | AY050440.1 | - | 1 - 2 | 442 | 154 | |
| BT020408.1 | - | 1 - 2 | 335 | 154 | ||
More Information
Set Membership: |
|

SPX4
Genomic Map