Assay Details
| Gene Symbol: LNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121562.3 | NP_197062.1 | 4 - 5 | 2566 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LNG1| Gene Symbol: LNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121562.3 | NP_197062.1 | 4 - 5 | 2566 | 70 | |
Set Membership: |
|