Assay Details
| Gene Symbol: AT5G15680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121572.5 | NP_197072.3 | 10 - 11 | 1182 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G15680| Gene Symbol: AT5G15680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121572.5 | NP_197072.3 | 10 - 11 | 1182 | 106 | |
Set Membership: |
|